Stable Cells
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage Conditions:LN₂ (Below –135 ℃)
Specifications
Related Information
MULTISCREEN Stable Cell Lines Expressing GPCRs and Beyond
Application Notes
Multispan MULTISCREEN™ Products and Services
Product Literature
TDS Beta-Arrestin2 Stable Cell Line Rat Recombinant CB2 Receptor (High Expression) TDS Beta-Arrestin2 Stable Cell Line Rat Recombinant CB2 Receptor (Low Expression) TDS Beta-Arrestin2 Stable Cell Line Rat Recombinant GPR40 Receptor TDS Human Recombinant ZNT3 Receptor TDS Rat Recombinant P2Y12 Receptor
Datasheet
More
Product Information
- Catalog #Supplier #Cell FormatSingle Donor/PooledSizeAvailabilityPrice
- MULTISCREEN™ Rat CB2 CHO-K1 β-Arrestin2 HTS Stable Cell LineMULTISCREEN™ Rat CB2 CHO-K1 β-Arrestin2 HTS Stable Cell Line76918-502CAR1230BA2-1ACryopreservedSingle donor2 VialsLog in for availabilityEach$48,435.75Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog 5-HT2A CHO-K1 HTS Stable Cell LineMULTISCREEN™ Dog 5-HT2A CHO-K1 HTS Stable Cell Line76919-058CD1324CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat CB2 CHO-K1 β-Arrestin2 HTS Stable Cell LineMULTISCREEN™ Rat CB2 CHO-K1 β-Arrestin2 HTS Stable Cell Line76918-750CAR1230BA2-1CryopreservedSingle donor2 VialsLog in for availabilityEach$48,435.75Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GIP HEK293T HTS Stable Cell LineMULTISCREEN™ Dog GIP HEK293T HTS Stable Cell Line76918-268CD1290CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR40 CHO-K1 β-Arrestin2 HTS Stable Cell LineMULTISCREEN™ Rat GPR40 CHO-K1 β-Arrestin2 HTS Stable Cell Line76918-622CAR1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$48,435.75Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno GPR40 CHO-K1 β-Arrestin2 HTS Stable Cell LineMULTISCREEN™ Cyno GPR40 CHO-K1 β-Arrestin2 HTS Stable Cell Line76918-980CAPC1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$48,435.75Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rhesus B2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rhesus B2 HEK293T HTS Stable Cell Line76918-372CP1199CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno GPR43 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Cyno GPR43 CHO-K1 HTS Stable Cell Line76919-370CPC1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog mGluR4 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Dog mGluR4 HEK293T Gαqi5 HTS Stable Cell Line76918-560HGD1191CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat TrKA CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat TrKA CHO-K1 HTS Stable Cell Line76919-192CKR1001-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno Glucagon HEK293T HTS Stable Cell LineMULTISCREEN™ Cyno Glucagon HEK293T HTS Stable Cell Line76919-108CPC1266CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno Glucagon CHO-K1 HTS Stable Cell LineMULTISCREEN™ Cyno Glucagon CHO-K1 HTS Stable Cell Line76918-944CPC1266-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat BB1 C6 HTS Stable Cell LineMULTISCREEN™ Rat BB1 C6 HTS Stable Cell Line76918-652C1211-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat UT CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat UT CHO dhfr- HTS Stable Cell Line76919-348C1035-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit B2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit B2 HEK293T HTS Stable Cell Line76919-230CB1199CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit GIP HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit GIP HEK293T HTS Stable Cell Line76918-466CB1290CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat KiSS1 CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat KiSS1 CHO dhfr- HTS Stable Cell Line76919-294C1036-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Human ZnT3 HeLa HTS Stable Cell LineMULTISCREEN™ Human ZnT3 HeLa HTS Stable Cell Line76918-368C2001-7CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR120 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GPR120 HEK293T HTS Stable Cell Line76919-026CR1294CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GIP HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GIP HEK293T HTS Stable Cell Line76918-342CR1290CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat Y2 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat Y2 CHO-K1 HTS Stable Cell Line76919-110CR1274-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno Glucagon CHO-K1 HTS Stable Cell LineMULTISCREEN™ Cyno Glucagon CHO-K1 HTS Stable Cell Line76918-706CPC1266-1ACryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Guinea Pig B2 HEK293T HTS Stable Cell LineMULTISCREEN™ Guinea Pig B2 HEK293T HTS Stable Cell Line76918-984CC1199CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GLP-2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GLP-2 HEK293T HTS Stable Cell Line76918-698CR1268CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog V2 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Dog V2 CHO-K1 HTS Stable Cell Line76919-326CD1044-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno GLP-1 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Cyno GLP-1 CHO-K1 HTS Stable Cell Line76919-144CPC1267-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GLP-1 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat GLP-1 CHO-K1 HTS Stable Cell Line76919-236CR1267-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit OT HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit OT HEK293T HTS Stable Cell Line76918-378CB1299CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rhesus BLT2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rhesus BLT2 HEK293T HTS Stable Cell Line76918-916CPR1272CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog B2 HEK293T HTS Stable Cell LineMULTISCREEN™ Dog B2 HEK293T HTS Stable Cell Line76919-150CD1199CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rhesus BLT2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rhesus BLT2 HEK293T HTS Stable Cell Line76918-276CPR1272ACryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Cyno GLP-1 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Cyno GLP-1 CHO-K1 HTS Stable Cell Line76918-426CPC1267-1ACryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GPR40 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Dog GPR40 CHO-K1 HTS Stable Cell Line76919-010CD1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GPR43 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Dog GPR43 CHO-K1 HTS Stable Cell Line76919-164CD1104-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rhesus GPR43 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rhesus GPR43 CHO-K1 HTS Stable Cell Line76919-368CPR1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat S1P2 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat S1P2 CHO-K1 HTS Stable Cell Line76918-428CR1051-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR40 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GPR40 HEK293T HTS Stable Cell Line76918-710CR1101CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat MCH1 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat MCH1 HEK293T Gαqi5 HTS Stable Cell Line76918-260C1031-4CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rhesus GIP HEK293T HTS Stable Cell LineMULTISCREEN™ Rhesus GIP HEK293T HTS Stable Cell Line76918-258CPR1290CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GLP-1 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Dog GLP-1 CHO-K1 HTS Stable Cell Line76918-660CD1267-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat LPA1 RH7777 HTS Stable Cell LineMULTISCREEN™ Rat LPA1 RH7777 HTS Stable Cell Line76918-266CR1048-6BCryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR40 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat GPR40 CHO-K1 HTS Stable Cell Line76918-574CR1101-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPBA CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat GPBA CHO-K1 HTS Stable Cell Line76919-118CR1361-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat Kappa Opioid CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat Kappa Opioid CHO dhfr- HTS Stable Cell Line76919-006CR1352-8CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat Mu Opioid CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat Mu Opioid CHO dhfr- HTS Stable Cell Line76918-686CR1350-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat NK3 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat NK3 HEK293T HTS Stable Cell Line76918-674CR1305CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat mGluR2 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat mGluR2 HEK293T Gαqi5 HTS Stable Cell Line76919-112HGR1189CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat A1 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat A1 HEK293T HTS Stable Cell Line76919-256CR1427CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat P2Y2 1321N1 HTS Stable Cell LineMULTISCREEN™ Rat P2Y2 1321N1 HTS Stable Cell Line76919-152CR1161-3CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat A2A HEK293T HTS Stable Cell LineMULTISCREEN™ Rat A2A HEK293T HTS Stable Cell Line76918-966CR1428CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat mGluR4 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat mGluR4 HEK293T HTS Stable Cell Line76918-800CR1191CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR91 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat GPR91 CHO-K1 HTS Stable Cell Line76918-538CR1144-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog CCR9 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Dog CCR9 HEK293T Gαqi5 HTS Stable Cell Line76918-950CGD1017CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GPR35 HEK293T Gα16 HTS Stable Cell LineMULTISCREEN™ Dog GPR35 HEK293T Gα16 HTS Stable Cell Line76919-296CGD1096CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat TSH HEK293T HTS Stable Cell LineMULTISCREEN™ Rat TSH HEK293T HTS Stable Cell Line76919-346C1177-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit V2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit V2 HEK293T HTS Stable Cell Line76918-730CB1044LCryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit V2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit V2 HEK293T HTS Stable Cell Line76918-328CB1044CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit V1B HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit V1B HEK293T HTS Stable Cell Line76918-792CB1043CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rabbit V1A HEK293T HTS Stable Cell LineMULTISCREEN™ Rabbit V1A HEK293T HTS Stable Cell Line76918-726CB1042CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat ETA CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat ETA CHO-K1 HTS Stable Cell Line76919-120CR1216-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat B2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat B2 HEK293T HTS Stable Cell Line76919-276CR1199CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat EP2 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat EP2 HEK293T HTS Stable Cell Line76919-136CR1202CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Ferret GIP HEK293T HTS Stable Cell LineMULTISCREEN™ Ferret GIP HEK293T HTS Stable Cell Line76919-188CF1290CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat P2Y14 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat P2Y14 HEK293T Gαqi5 HTS Stable Cell Line76919-130CGR1057CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat TA1 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat TA1 HEK293T HTS Stable Cell Line76919-316C1357CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat S1P4 CHO-K1 Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat S1P4 CHO-K1 Gαqi5 HTS Stable Cell Line76919-218CGR1052-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat CCR9 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat CCR9 HEK293T Gαqi5 HTS Stable Cell Line76918-866CGR1017CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Monkey P2Y14 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Monkey P2Y14 HEK293T Gαqi5 HTS Stable Cell Line76918-576CGP1057CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR17 1321N1 HTS Stable Cell LineMULTISCREEN™ Rat GPR17 1321N1 HTS Stable Cell Line76918-658CR1526-3CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat NOP CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat NOP CHO dhfr- HTS Stable Cell Line76918-554C1354CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog GPR35 CHO-K1 Gα16 HTS Stable Cell LineMULTISCREEN™ Dog GPR35 CHO-K1 Gα16 HTS Stable Cell Line76918-534CGD1096-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR88 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat GPR88 CHO-K1 HTS Stable Cell Line76918-492CR1141-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat A2B HEK293T HTS Stable Cell LineMULTISCREEN™ Rat A2B HEK293T HTS Stable Cell Line76918-678CR1429CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR43 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GPR43 HEK293T HTS Stable Cell Line76919-020CR1104CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat GPR41 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat GPR41 HEK293T HTS Stable Cell Line76918-334CR1102CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat A3 HEK293T HTS Stable Cell LineMULTISCREEN™ Rat A3 HEK293T HTS Stable Cell Line76918-350CR1430CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog mGluR4 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Dog mGluR4 HEK293T Gαqi5 HTS Stable Cell Line76918-464CGD1191CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat NPBW1 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat NPBW1 HEK293T Gαqi5 HTS Stable Cell Line76918-346C1124-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat P2Y12 CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat P2Y12 CHO dhfr- HTS Stable Cell Line76918-474C1170-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Dog mGluR4 CHO-K1 Gαqi5 HTS Stable Cell LineMULTISCREEN™ Dog mGluR4 CHO-K1 Gαqi5 HTS Stable Cell Line76918-476CGD1191-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat Delta Opioid CHO dhfr- HTS Stable Cell LineMULTISCREEN™ Rat Delta Opioid CHO dhfr- HTS Stable Cell Line76919-314C1351CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat S1P3 CHO-K1 Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat S1P3 CHO-K1 Gαqi5 HTS Stable Cell Line76919-372CGM1049-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat CB2 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat CB2 CHO-K1 HTS Stable Cell Line76919-116CR1230-1ACryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat CaSR HEK293T HTS Stable Cell LineMULTISCREEN™ Rat CaSR HEK293T HTS Stable Cell Line76919-096CR1233CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat MT2 HEK293T Gαqi5 HTS Stable Cell LineMULTISCREEN™ Rat MT2 HEK293T Gαqi5 HTS Stable Cell Line76918-518CR1224CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- MULTISCREEN™ Rat CB2 CHO-K1 HTS Stable Cell LineMULTISCREEN™ Rat CB2 CHO-K1 HTS Stable Cell Line76918-684CR1230-1CryopreservedSingle donor2 VialsLog in for availabilityEach$36,057.73Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.