Mouse Glycogen Phosphorylase MM,GP-MM ELISA Kit
76701-370
:
- Assay Type (ELISA with LOV):Sandwich
- Host:Mouse
- Target protein:Glycogen phosphorylase MM,GP-MM
- Description:Mouse Glycogen phosphorylase MM,GP-MM ELISA kit
- Size:96 wells
- Sample Type:Serum, plasma, urine, tissue homogenates, cell culture supernates
- Time to Results:1 - 5 h
- Shelf Life:6 months (4 °C); 12 months (−20 °C)
- Storage Temperature:2…8 °C
- Sample Volume:50 - 100 μl
- Sensitivity:1 - 5 h
- Regulatory Status:RUO
- Cat. No.:76701-370
- Detection wavelength: 450 nm