CD4 is glycoprotein found on the surface of immune cells such as T helper cells, monocytes, macrophages, and dendritic cells. It is a co-receptor that assists the T-cell receptor (TCR) with antigen-presenting cell and also interacts directly with MHC class II molecules of the surface of antigen presenting cells. Most mature T-cell lymphomas are CD4 positive with the exception of aggressive NK-cell leukemia and extranodal NL/T-cell lymphoma. CD4 plays an important role in the classification of lymphocytes in inflammatory legions and malignant lymphomas.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76634-806
- 76634-804
- Antigen Name:
- CD4
- CD4
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- EP204
- EP204
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Size
Specifications
Antigen NameCD4Environmentally PreferableAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneEP204ReactivityHumanHostRabbitIsotypeIgGSpecifications
Antigen NameCD4Environmentally PreferableAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneEP204ReactivityHumanHostRabbitIsotypeIgG