Inhibin is a dimeric glycoprotein hormone from the TGF-β family made of β and α subunits. This antibody recognizes the 32kDa α subunit of human inhibin. Inhibin α is expressed in a wide variety of human tissues, such as brain, prostate, adrenal, granulose cells of ovary, sertoli cells of testis and in fetoplacental unit.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 76633-904
- 76633-908
- 76633-906
- Antigen Name:
- Inhibin
- Inhibin
- Inhibin
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- R1
- R1
- R1
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- IgG2a
- Size
Specifications
Antigen NameInhibinEnvironmentally PreferableAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneR1ReactivityHumanHostMouseIsotypeIgG2aSpecifications
Antigen NameInhibinEnvironmentally PreferableAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneR1ReactivityHumanHostMouseIsotypeIgG2aSpecifications
Antigen NameInhibinEnvironmentally PreferableAntibody TypePrimaryClonalityMonoclonalConjugationUnconjugatedCloneR1ReactivityHumanHostMouseIsotypeIgG2a