20LFLHE Tychem® hoods operate with the entire Bullard® PAPR family to offer respiratory protection and filter out exhaled air through a HEPA panel.
- Constructed with Tychem® 2000 and 99.97% HEPA filtration panel
- Non-latex sealing surface provides maximum comfort
- No fit testing required
- Over the top airflow keeps operator cool and comfortable
- Built-in suspension system
- Assigned protection factor of 25
The 20LFHE series hood offers unique customer solutions by providing outstanding, comfortable respiratory protection as well as filtering the operators exhaled air through HEPA filtration panel located in base of the hood. Positive pressure airflow is much more comfortable than tight fitting masks that require costly fit testing. Available in both standard fit and narrow profile, which provides a snugger fit.
Certificaciones: NIOSH certified with Bullard® PAPR systems.