Thermo-Dynamic steam trap for clean applications
Supplier: Spirax Sarco
BTD52L 1810298
MFLX07600-24EA 1164 USD
MFLX07600-24
Thermo-Dynamic steam trap for clean applications
Filters Gas Filters
Keep steam space free of condensate to provide maximum system efficiency.
  • Constructed of durable 316L stainless steel (SS)
Made of 316L stainless steel for aggressive condensate found in clean steam systems.
Typical applications:
Biotech, pharmaceutical, fine chemical
Food, dairy, and beverage industries
WFI system sterilization
Drainage of steam filter housing and separators