Stainless Steel Multiple Filter Housings
Supplier: Pentair
156025-02
MFLX29810-05EA 1893.75 USD
MFLX29810-05
Stainless Steel Multiple Filter Housings
Filters Filter Cartridges
Stack up to 20 double open end cartridges in one housing!
  • Use standard double-open-end cartridges for high-flow applications
  • Easy to install and maintain with wide-top access port
Stainless steel housings are great for drinking water applications in schools, restaurants, institutions, and industrial plants. Housings hold from 4 to 20 double open end cartridges--filter a large volume of water every minute! Housings feature durable 304 stainless steel construction with epoxy finish to provide better chemical compatibility. All ports are clearly labeled for easy installation.
Maximum pressure is 125 psi; maximum temperature is 300 °F (149 °C). Clean and dirty drains are 1/4", inlet and outlet ports are 2" NPT (M). Order cartridges separately. Buna N seal.