Valeryl fentanyl-13C6 (CRM), Cayman Chemical
76544-440
:
- Concentration:100 µg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- Description:Valeryl fentanyl-13C6 (CRM)
- Size:100 µg
- Synonym(s):Pentanoyl fentanyl-13C6; Pentanyl fentanyl-13C6
- Formula:C₁₈[₁₃C]₆H₃₂N₂O
- Storage Temperature:–20 °C
- Description:Valeryl fentanyl-13C6
- Cat. No.:76544-440
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard