Benchmark Hotplate
Supplier: Benchmark Scientific
H4000-H
490005-864EA 488.98 USD
490005-864
Benchmark Hotplate
Heaters Hot Plates
Hotplate that is compatible with a wide variety of popular sizes of borosilicate glass beakers, flasks and bottles.
  • Large white ceramic top plate
  • Compact, 8x9" footprint
  • Safety indicator LED's clearly indicate hot surface
These hotplates feature an exceptionally durable, chemical resistant white ceramic work surface.
Advanced microprocessor controls with convenient turn knob allow quick, precise adjustment and maintenance of temperature.
The hotplate comes complete with a support rod for mounting thermometers and temperature probes.
Their space-efficient design makes them ideal for use on crowded benchtops and inside of bio-hoods and incubators.
Corresponding magnetic hotplate stirrers (heating and stirring 490005-866) or magnetic stirrers (stirring only 490005-868) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Información para pedidos: Two-year warranty.