


Oxalic acid, anhydrous 98%
:144-62-7
:
:C₂H₂O₄
:MFCD00002573
:90.04 g/mol
:365 °C (1013 hPa)
:189…191 °C
:1.9 g/cm³
:>93.4 °C
:GHS05
:GHS07
:GHS08
:Ambient
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:189 to 191 °C
- Titration with NaOH:97.5 to 102.5 %
- pH:8.5 to 10.5 (10 % suspension in water)
- Pack typeSize
- TS18643-250018643-2500Glass bottle for solids250 gEach (250g)$56.00
- TS18643-001018643-0010Glass bottle for solids1 kgEach (1kg)$126.00