Puerto Rico
Order Entry
Pyrene 98%
 
Pyrene 98%
 
undefined
 
Pyrene 98%

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Appearance:
    Light yellow to yellow to yellow-green Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    147 to 152 °C
  • GC:
    ≥97.5 %

 

 

  •  
     
    Pack type
    Size
     
     
  • TS18083-0250
    18083-0250
    Glass bottle for solids
    25 g
       
       
    Each (25g)
    Each (25g) - $38.00
    $38.00
    Each (25g)
       $38.00

  • TS18083-1000
    18083-1000
    Glass bottle
    100 g
       
       
    Each (100g)
    Each (100g) - $90.00
    $90.00
    Each (100g)
       $90.00

  • TS18083-5000
    18083-5000
    Glass bottle
    500 g
       
       
    Each (500g)
    Each (500g) - $293.00
    $293.00
    Each (500g)
       $293.00