VWR® Critical Environment Swab, Small Foam Head with Flexible Tip, VWR International
Supplier: VWR, part of Avantor
VWR®
SU-068VFCTD
76478-854BX 101.52 USD
76478-854
VWR® Critical Environment Swab, Small Foam Head with Flexible Tip, VWR International
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • Small size head
Swab head is thermally bonded to the handle without using any adhesive.