VWR® Critical Environment Swab, Mini Pointed Compressed Foam Head, VWR International
Supplier: VWR, part of Avantor
VWR®
SU-076VFSTD
76478-850BX 100.18 USD
76478-850
VWR® Critical Environment Swab, Mini Pointed Compressed Foam Head, VWR International
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • Mini pointed foam head
Swab head is thermally bonded to the handle without using any adhesive.