VWR® Critical Environment Swab, Medium Foam Head
Supplier: Avantor
VWR®
SU-162VFCTD
76478-844BX 184.3 USD
76478-844
VWR® Critical Environment Swab, Medium Foam Head
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane foam with a long, 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Polypropylene handle
  • Medium size head
  • No adhesive
Swab head is thermally bonded to the handle without using any adhesive.