VWR® Critical Environment Swab, Medium Flexible Foam Head, VWR International
Supplier: VWR, part of Avantor
VWR®
SU-103VFCTD
76478-840BX 40.51 USD
76478-840
VWR® Critical Environment Swab, Medium Flexible Foam Head, VWR International
Swabs Swabs for Clinical Use
Swab features 100 ppi closed cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam swab head
  • Polypropylene handle
  • Medium size head
Swab head is thermally bonded to the handle and augmented with adhesive.