Nitrogen Generator, Infinity XE 5011, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
3300189
103549-808EA 21581.54 USD
103549-808
Nitrogen Generator, Infinity XE 5011, Peak Scientific
Gas Generators Nitrogen Generators
Producing between 10-130 L/min of high quality, laboratory grade nitrogen gas, the Infinity XE 501X can comfortably supply gas for multiple laboratory instruments.
  • Flow rates from 10-130 L/min
  • Variable flow rate, with up to 135 psi outlet pressure
  • N2 purity from 95-99.5% (dependent on flow rate)
  • Tried and tested membrane technology for high quality gas
  • Small footprint using minimal laboratory space
  • Effectively silent in operation
  • O₂ analyzer option available, XE 5011 model includes PurityGuard for purity critical applications
Producing between 10-130 L/min of high quality, laboratory grade nitrogen gas, the Infinity XE 501X can comfortably supply gas for multiple laboratory instruments. With variable nitrogen purity available between 95-99.5% (depending on flow rate), Infinity XE generators can be configured to suit the needs of several applications, such as LC-MS, ELSD, sample preparation and more.
Peak Scientific's Infinity XE series generators have been engineered to provide nitrogen gas to laboratories that already have an external source of clean, dry and oil-free air; and meets a minimum grade of ISO8573-1:2010 Class 1.4.1 - NB: Were this is not available, Peak can provide consultative assistance on what specifications of compressed air and pre-filtration are required, as well as supply these items as part of a complete Infinity XE gas generation solution.
Infinity XE series nitrogen generators are effectively silent whilst in operation and have no moving parts, so require minimal annual maintenance.
Models available with or without optional O₂ analyzer and purity safeguard system.