Nitrogen Generator, NG3000A ultra high purity, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
10-1230
103549-812EA 14547.69 USD
103549-812
Nitrogen Generator, NG3000A ultra high purity, Peak Scientific
Gas Generators Nitrogen Generators
The NG3000A delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of 3 L/min for GC, DSC, DMA and other applications requiring ultra high purity nitrogen at up to 3L/min.
  • Suitable for applications such as GC, DSC, DMA
  • Suitable for applications requiring ultra high purity
  • Available with or without an external compressor
  • Contains self-regenerating Carbon Molecular Sieve column
  • Gas is supplied on demand so generator works to your schedule
  • 12 month comprehensive on-site warranty
The NG3000A delivers ultra high purity nitrogen at trace detection limits through Pressure Swing Adsorption technology at flows of 3 L/min for GC, DSC, DMA and other applications requiring ultra high purity nitrogen at up to 3L/min. The NG3000A comes with an internal compressor for laboratories without an external source of gas.
To reach their ultra high purity nitrogen output these generators use Pressure Swing Adsorption and Carbon Molecular Sieve. These separation technologies remove oxygen and other impurities in air, delivering hassle-free, ultra high purity nitrogen, on-demand for the laboratory.