VWR® Performance Touch Screen Glass Door Laboratory Refrigerators
Supplier: VWR, part of Avantor
VWR®
HCLP-72-TS HCLP-49-TS HCLP-19-TS HCLP-26-TS
10819-542EA 17563.67 USD
10819-542 10820-344 10819-538 10819-534
VWR® Performance Touch Screen Glass Door Laboratory Refrigerators
Refrigerators
These performance glass door refrigerators offer top-quality features in laboratory cold storage with advanced data logging and superior temperature control.
  • Touch screen digital temperature display and alarm module
  • Extensive alarms including audible, visual, senor error, power failure, low battery, and door ajar alarms
  • Temperature data logging and USB port for data transfer and software updates
  • Microprocessor temperature controller for precise temperature regulation
  • Keyed door lock, LED interior lighting, and adjustable shelving
  • Natural, hydrocarbon refrigerant
These high-performance laboratory refrigerators are designed with extensive unit alarms, touch-screen displays, and advanced capabilities for temperature data logging and transfer. All units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a three year parts and labor warranty, plus an additional two year compressor parts warranty.