VWR® Standard Hazardous Location (Explosion Proof) Refrigerators with Natural Refrigerants
Supplier: Avantor
VWR®
HCERV-17 HCERV-20 HCERV-14
76499-002EA 6143.39 USD
76499-002 76499-000 76499-004
VWR® Standard Hazardous Location (Explosion Proof) Refrigerators with Natural Refrigerants
Refrigerators
These refrigerators are engineered for cold-storage use in environments where volatile conditions could potentially exist.
  • Microprocessor temperature controller
  • High density urethane foam cabinet and door insulation
  • Adjustable shelving
  • Keyed door lock
  • Natural, hydrocarbon refrigerant (R600a)
These refrigerators are equipped with keyed door locks, interior shelving, and microprocessor temperature controllers. All units include a two year parts and labor warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certificaciones: ETL Listed. These units meet NFPA and OSHA guidelines Class 1, Division II, Group C & D for storing substances in hazardous locations.
Precaución: Explosion Proof Units are designed for environments where external hazards could exists. For storage of volatile substances in normal laboratory conditions, see VWR® Flammable Storage Units.