VWR® Standard Laboratory Refrigerators with Natural Refrigerants
Supplier: VWR, part of Avantor
VWR®
HCRFS-20 HCRFS-17 HCRFS-14
76498-990EA 4939.97 USD
76498-990 76498-988 76498-986
VWR® Standard Laboratory Refrigerators with Natural Refrigerants
Refrigerators
These standard laboratory refrigerators are economical and multi-functional, performing well in a variety of laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant (R290)
All units feature a series of temperature alarms to keep stored products at peak effectiveness. These refrigerators are equipped with keyed door locks, interior shelving, probe access, and microprocessor temperature controllers.
All units include a two year parts and labor warranty. The refrigerators run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certificaciones: ETL Listed.