Orum International Stainless Steel Satellite with HEPA Filter, contact plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS263K BAS262K
76470-204EA 3635.38 USD
76470-204 76470-202
Orum International Stainless Steel Satellite with HEPA Filter, contact plate, Hardy Diagnostics
Samplers Air Samplers
Accessory option for ISOLATOR RABS (76377-520, 76377-522, 76377-524, 76377-526) instrument for facilities requiring HEPA-equipped microbial air samplers.
Información para pedidos: When using the HEPA satellite with the TRIO.BAS™ RABS ISOLATOR, all satellites must be of the same catalog number. The HEPA satellite format cannot be used concurrently with standard, or other, satellite configurations.