You Searched For: Traffic Cones/Barriers

Reducing accidents and liabilities, the traffic cones and barriers keep individuals from entering hazardous areas. Offered in bright colors for high visibility, the products are solutions to dangerous occupations by providing additional safety to workers that would otherwise be vulnerable. Traffic cones and barriers feature universal graphics to efficiently communicate potential risks regardless of the spoken languages. With weather resistance, the versatile chains, paddle signs, and cones can be used indoors or outdoors.

Reducing accidents and liabilities, the traffic cones and barriers keep individuals from entering hazardous areas. Offered in bright colors for high visibility, the products are solutions to dangerous occupations by providing additional safety to workers that would otherwise be vulnerable. Traffic cones and barriers feature universal graphics to efficiently communicate potential risks regardless of the spoken languages. With weather resistance, the versatile chains, paddle signs, and cones can be used indoors or outdoors.


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (56620-804)
Supplier: New Pig
Description: The 0.125"-thick polyethylene shell resists impact, chemicals and UV light


Supplier: National Marker
Description: STOP/ SLOW paddle signs are used during road construction to help guide traffic through one way roadways

Supplier: ACCUFORM MANUFACTURING, INC
Description: Outdoor Retractable Belt Tape Barriers direct the flow of pedestrian traffic.

Supplier: ACCUFORM MANUFACTURING, INC
Description: BLOCKADE Retractable Belt Tape Barriers direct the flow of pedestrian traffic inside any building or facility.

Supplier: ACCUFORM MANUFACTURING, INC
Description: Bucket of plastic chains allows you to set up a barrier quickly and economically when used with stanchion posts (sold separately).

Supplier: National Marker
Description: For use with plastic chain.

Supplier: National Marker
Description: For use with plastic chain.

Supplier: ACCUFORM MANUFACTURING, INC
Description: Plastic chains allows you to set up a barrier quickly and economically when used with stanchion posts (sold separately.)

Supplier: Janitorial Supplies
Description: The Rubbermaid Commercial pop-up safety cone with multi-language imprint and wet floor symbol is collapsible and automatically deployed.

Supplier: ACCUFORM MANUFACTURING, INC
Description: Outdoor Retractable Belt Tape Barriers direct the flow of pedestrian traffic.

Supplier: National Marker
Description: Hang chains between a series of warning posts or barrels for crowd control or guiding traffic

Supplier: National Marker
Description: Warn of potential hazards from three sizes

Supplier: National Marker
Description: Used to attach lengths of chain.

Supplier: National Marker
Description: Get double duty out of your traffic cones

Catalog Number: (76442-598)
Supplier: ACCUFORM MANUFACTURING, INC
Description: BLOCKADE Retractable Belt Tape Barriers direct the flow of pedestrian traffic inside any building or facility, and can be used to remind pedestrians t...


Catalog Number: (76471-370)
Supplier: ACCUFORM MANUFACTURING, INC
Description: BLOCKADE Retractable Belt Tape Barrier Receiver Posts are designed for use at the end of an extended Belt Tape Barrier.


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom