You Searched For: Hematology Stains
Hematology stains and reagents are essential for clinical and medical cytology research. Hard to see blood, bone marrow, or lymphatic system components is easily seen and identified by staining. Whether manually or automatically processing, the Wright, Giesma, and Leishman formulations work efficiently on prepared blood smears. Working with time sensitive samples, all the hematology stains are designed to work rapidly for the most accurate results. Romanowsky stain solutions are used in hematology. They are composed of methylene blue, oxidative products of methylene blue (Azure A, Azure B, Azure C, and Thionin) and eosin dyes. Giemsa, a commonly used stain, does not adequately stain red blood cells, platelets, or white blood cell cytoplasms when used alone.
Hematology stains and reagents are essential for clinical and medical cytology research. Hard to see blood, bone marrow, or lymphatic system components is easily seen and identified by staining. Whether manually or automatically processing, the Wright, Giesma, and Leishman formulations work efficiently on prepared blood smears. Working with time sensitive samples, all the hematology stains are designed to work rapidly for the most accurate results. Romanowsky stain solutions are used in hematology. They are composed of methylene blue, oxidative products of methylene blue (Azure A, Azure B, Azure C, and Thionin) and eosin dyes. Giemsa, a commonly used stain, does not adequately stain red blood cells, platelets, or white blood cell cytoplasms when used alone.
14  results were found
SearchResultCount:"14"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR International
Description: EASY III™ is a three step, rapid differential stain.
Supplier: Epredia
Description: Rapidly and conveniently stain blood smears with Epredia™ Buffered Wright Stain, a single-solution stain that contains fixative, buffer and stain.
SDS
Catalog Number: (76422-822)
Supplier: Hardy Diagnostics
Description: A Wrights-Giemsa stain, buffer, and rinse for use with the QuickSlide™ HemaPRO automated hematology slide stainer.
Supplier: Azer Scientific Inc.
Description: Hematology stain packs are designed for use in Hema-Tek™ I, II, 1000, and 2000 model bench top automated stainers.
Supplier: Electron Microscopy Sciences
Description: Carstairs Method for Fibrin and Platelets is used for the staining of fibrin and platelets
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: Giemsa Thick Film Stain, Lillie (1965); Barber and Komp (1929), Stain malarial parasites, Stain results - Malarial Parasites: Clear Red Chromatin, Cytoplasm: Clear Blue, Red Corpuscles: Not seen due to lacking of
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: Neat Stain™ Kits are specifically designed for stat procedures and/or laboratories using manual staining methods. Neat Stain™ is available as a rapid ...
Supplier: Electron Microscopy Sciences
Description: Hall’s Method for Bilirubin
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: May-Grunwald / Giemsa Combined Stain, Beck (1938); Pappanheim (1912), Stain results - Nuclei, Bacteria Blue, Cytoplasm Pink-Rose
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: This kit is a Modified Giemsa Stain kit for quick turnaround results. It may be used for rapid blood smears for differential assessment, as well as fo...
Supplier: Electron Microscopy Sciences
Description: 3-Amino-9-Ethyl Carbazole Method for Leukocyte Peroxidase, Stains for hematologic elements, Stain results - Sites of Peroxidase activity: Red-Brown Granules, Nuclei: Blue
Minority or Woman-Owned Business Enterprise
Supplier: Electron Microscopy Sciences
Description: Differentiation of leucocytes, rickettsia, bacteria and inclusion bodies, Stain results - Nuclei, additional leukocyte features are similar to, Wright Stain results - Reddish Purple, Plasmodic cytoplasm: Blue
Minority or Woman-Owned Business Enterprise
Supplier: ELITECHGROUP INC. MS
Description: Reagents designed for staining peripheral blood smears with the aerospray hematology slide stainer.
Catalog Number: (103783-058)
Supplier: IMEB INC MS
Description: The Tek-Select® Diff-Stain kit is designed to provide Nuclear and cytoplasmic staining and recently has provided excellent staining for Helico...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 14 of 14
no targeter for Bottom