You Searched For: Parasitology Containers
Sealed, break-resistant parasitology containers are used for the collection, transport, and examination of fecal and urine samples. Concentrators can be used to remove certain preparation steps and provide an enclosed system for lab personnel so there is no exposure to stool or exposed fumes. Fixatives and preservatives are available in separate kits for some models to meet individual laboratory needs. Enhanced filtration methods improve sample clarity and reduce the amount of debris on slides, making them easier to examine.
8 results were found
Sort Results
List View
Easy View