Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Hematology Controls"

https://pr.vwr.com/stibo/smallweb/std.lang.all/93/94/18989394.jpg

The hematology controls contain blood parameters at low, normal, or high levels to monitor equipment performance and therefore test precision. By routinely checking the analytic linearity of systems used, the chance of inaccurate patient test results being reported is significantly minimized. Compatible with a variety of analyzers, the versatile tubes are ready-to-run and act identically to human supplied samples. The hematology controls have extended shelf time storage capabilities for long term stability.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
Sort by:
 
 
 
 

Supplier:  Thermo Scientific
Description:   DATA-TROL A (ABNORMAL)
Supplier:  Thermo Scientific
Description:   Pacific Hemostasis Abnormal Coagulation Reference Plasma (ACRP) 1ml
SDS
Supplier:  NERL Diagnostics
Description:   Pacific Hemostasis Reference Emulsion Fluid 500ml
SDS
Supplier:  Thermo Scientific
Description:   Pacific Hemostasis High Fibrinogen Control 1ml
SDS
Supplier:  Promocell
Description:   Calpain 1 Inhibitor Screening Kit
Supplier:  Wako Life Sciences
Description:   Control Serum Combo
Catalog Number: (97000-004)
Supplier:  Wako Life Sciences
Description:   Autokit Micro Albumin
Supplier:  Polysciences Inc
Description:   PROTEIN A-FLUOREBRITE YG
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next