You Searched For: Solution Bottles

Solution bottles are designed for storing liquids or media for preparation. While smaller sizes follow conventional shaping, larger sizes being similar in design to carboys. Solution bottles are available plain or with graduations for easy measuring. Necks are uniquely tooled for increased mechanical strength and for a uniform fit with rubber stoppers. All large-sized heavy-duty solution bottles meet federal specifications for quality and safety.

Solution bottles are designed for storing liquids or media for preparation. While smaller sizes follow conventional shaping, larger sizes being similar in design to carboys. Solution bottles are available plain or with graduations for easy measuring. Necks are uniquely tooled for increased mechanical strength and for a uniform fit with rubber stoppers. All large-sized heavy-duty solution bottles meet federal specifications for quality and safety.


16  results were found

SearchResultCount:"16"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: We offer a big variety of packaging materials like empty bottles and containers for your own preparations and mixtures of our reagents.

SDS

Supplier: DWK Life Sciences (KIMBLE)
Description: Designed to store and discharge liquids via a bottom valve outlet.
Supplier: Corning
Description: PYREX® borosilicate glass.
Supplier: DWK Life Sciences (KIMBLE)
Description: Ideal for storage and dispensing of solutions.

Supplier: DWK Life Sciences (KIMBLE)
Description: Narrow mouth solution bottles for use with rubber stoppers.

Supplier: DWK Life Sciences (KIMBLE)
Description: KimCote® solution bottles are designed for storage of solutions and ideal for media preparations.

Supplier: Corning
Description: Designed for storage of solutions or media preparation. The 13.25 L sizes are conventional shape; the 19 and 45.5 L sizes are similar to carboys in de...
Catalog Number: (97021-550)
Supplier: Hach
Description: Soaker bottle for electrode short-term and long-term storage.

Product available on GSA Advantage®


Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Description: Narrow-mouth bottles conform to ASTM E438, Type 1, Class A standards.
Catalog Number: (89097-348)
Supplier: DWK Life Sciences (KIMBLE)
Description: KimCote® plastic safety coated solution bottles with carboy clamp and silicone stopper assemblies for solution bottles.


Supplier: Thermo Fisher Scientific
Description: For temporary disposal of most chemical and biological wastes. Each system consists of a waste container, removable HDPE funnel and cover, polypropyle...
Supplier: Foxx Life Sciences
Description: Borosilicate glass 3.3 bottles, with tooled neck, high mechanical strength and chemical resistant.

Supplier: DWK Life Sciences (KIMBLE)
Description: Borosilicate glass carboy-style bottles feature a narrow neck and sloping shoulders.
Supplier: Chemglass
Description: Complete reservoir system includes a plastic-coated graduated bottle, a standard cap assembly, solvent and sparging filters.

Small Business Enterprise

Catalog Number: (97050-590)
Supplier: DWK Life Sciences (KIMBLE)
Description: Glass solvent bottle is 1 L and has an 8 mm connector for HTS, HTS-xt, HTC, HTC-xt, HTX, and HTX-xt PAL systems.


Supplier: Chemglass
Description: Solvent reservoir for use in HPLC mobile phase delivery.

Small Business Enterprise

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 16
no targeter for Bottom