VWR® Plus Flammable Storage Refrigerator
Supplier: Avantor
FRS-30
10819-906EA 9511.63 USD
10819-906
VWR® Plus Flammable Storage Refrigerator
This VWR® Plus flammable storage refrigerator meets NFPA and OSHA guidelines 45 and 70 for the storage of flammable (volatile) materials in general laboratory work areas.
  • Microprocessor temperature controller
  • Digital temperature display
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (⅜")
  • CFC-free refrigerant
The flammable storage refrigerator is designed for laboratory use in compliance with NFPA and OSHA guidelines 45 and 70 for the storage of flammable materials. No electrical components exist in the interior of the unit and the compressor’s electrical components are sealed in a vapor-proof enclosure to reinforce safety.
Unit includes a series of temperature alarms for close temperature monitoring. Built with high-density foam insulation and shelves for product storage. Unit requires periodic defrosting to prevent ice build-up. Flammable storage units run on CFC-free refrigerant for economic efficiency and environmental safety.
Material: White textured cabinet and door
Units include a two year parts and labor warranty.
Certificaciones: Unit meets NFPA guidelines 45 and 70