VWR® Stem Thermometer, Calibrated, Electronic, Stainless Steel, 76 mm Probe
Supplier: VWR Life Science
VWR®
60900-3611
76472-448CS 547.33 USD
76472-448
VWR® Stem Thermometer, Calibrated, Electronic, Stainless Steel, 76 mm Probe
Thermometers Digital Thermometers
Digital stem thermometer is suitable for field studies, wet lab applications, wash down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialized, CE marked
Certificaciones: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Información de suministro: Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Precaución: These electronic stem thermometers are not suitable for solids such as frozen items.