C&A Scientific Flocked Swab for Oropharyngeal or Anterior Nares Sampling, Standard Tip, Hardy Diangostics
Supplier: Hardy Diagnostics
972029
76463-326EA 65.43 USD
76463-326
C&A Scientific Flocked Swab for Oropharyngeal or Anterior Nares Sampling, Standard Tip, Hardy Diangostics
Swabs Swabs for Sampling
Individually wrapped sterile swabs feature a flocked nylon tip and a breakpoint of 30 mm.
  • Anterior Nares swab (Oropharyngeal flocked swab)
  • Individually wrapped, medical grade paper
  • Sterile
  • ABS plastic handle
  • Flocked nylon tip
  • Scored breakpoint
  • FDA registered manufacturing facility
Swabs are individually wrapped in medical-grade paper peel-packaging and are designed for the collection of clinical samples. The swabs are sterilized via ethyl-oxide gas.