SP Wilmad-LabGlass Bulb for Pasteur Pipettes, SP Industries
Supplier: SP Industries
804
76459-578PK 29.9 USD
76459-578
SP Wilmad-LabGlass Bulb for Pasteur Pipettes, SP Industries
Pipets Pipet Filler Bulbs
Ideal for Pasteur pipettes used in NMR sample transfer.
  • Made from 100% latex
  • Manufactured in a clean room
Easily attached to all glass Pasteur pipettes.