Puerto Rico
Order Entry
Anti-Bevacizumab Hybridoma Polyclonal Antibody (For Neutralizing assay)
Anti-Bevacizumab Hybridoma Polyclonal Antibody (For Neutralizing assay)
 
Anti-Bevacizumab Hybridoma Polyclonal Antibody (For Neutralizing assay)
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 103815-438
  • 103815-440
  • Antigen Symbol:
  • Bevacizumab
  • Bevacizumab
  • Antigen Name:
  • Bevacizumab
  • Bevacizumab
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Powder
  • Powder
  • Purification:
  • Affinity purified
  • Affinity purified
  • Cross Adsorption:
  • Recognizes Bevacizumab specifically, no cross reactivity with other humanized antibodies.
  • Recognizes Bevacizumab specifically, no cross reactivity with other humanized antibodies.
  • Storage Temperature:
  • For long term storage, the product should be stored at lyophilized state at -20°C or lower.
  • For long term storage, the product should be stored at lyophilized state at -20°C or lower.
  • Shipping Temperature:
  • RT
  • RT

 

 

  •  
     
    Size
     
     
  • 103815-438
    BEBY121MG
    1 mg
       
       
    Each (1mg)
    Each (1mg) - $2,417.69
    $2,417.69
    Each (1mg)
       $2,417.69

     
     
     
    Specifications
    Antigen Symbol
    Bevacizumab
    Antigen Name
    Bevacizumab
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Isotype
    ELISA
    Yes
    Form
    Powder
    Purification
    Affinity purified
    Cross Adsorption
    Recognizes Bevacizumab specifically, no cross reactivity with other humanized antibodies.
    Storage Temperature
    For long term storage, the product should be stored at lyophilized state at -20°C or lower.
    Shipping Temperature
    RT
  • 103815-440
    BEBY12100UG
    100 ug
       
       
    Each (100µG)
    Each (100µG) - $333.85
    $333.85
    Each (100µG)
       $333.85

     
     
     
    Specifications
    Antigen Symbol
    Bevacizumab
    Antigen Name
    Bevacizumab
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Isotype
    ELISA
    Yes
    Form
    Powder
    Purification
    Affinity purified
    Cross Adsorption
    Recognizes Bevacizumab specifically, no cross reactivity with other humanized antibodies.
    Storage Temperature
    For long term storage, the product should be stored at lyophilized state at -20°C or lower.
    Shipping Temperature
    RT