Diversey™ Avert® Sporicidal Disinfectant Cleaner, Essendant
76461-964
- Description:Avert® Sporicidal Disinfectant Cleaner
- Size:12× 32 oz.
- Cat. No.:76461-964
- Packaging:Spray bottle
- Form:Liquid
Sporicidal disinfectant cleaner cleans, disinfects and deodorizes hard, nonporous surfaces in one labor saving step, with no rinsing required. Effective in 4 minutes against Clostridium difficile (C. diff) spores. Bleach-based formula eliminates quat-binding concerns. Designed for use on hard, nonporous surfaces such as those found in hospitals and other healthcare facilities.
- Cleans, disinfects, and deodorizes hard, nonporous inanimate (environmental) surfaces (and patient care equipment) in one step with no rinsing required
- Effective in 4 minutes against Clostridium difficile (C. diff) (C. difficile) spores
- Bleach-based formula means no quat-binding concerns
- Designed for use on hard, nonporous surfaces
- No added fragrances or dyes
Great for cleaning and disinfecting sinks, faucets, fixtures, bathrooms, restrooms, tubs, showers and more.
Application: Disinfectant/Sanitizer.
Applicable Material: Hard, Nonporous Surfaces.
Chemical Compound: Sodium Hypochlorite.