Gloves provide excellent physical durability when compared to conventional disposable gloves.
- Non sterile and Latex-free
- 3.5 g weight
- Minimum thickness of 2.0 mil
- Powder-free
- 9" length
- Medical grade
- Finger textured with beaded cuff
Certificaciones: ASTM D6124, ASTM D412/ ASTM D573, ASTM D3767, ASTM D6319. The gloves are manufactured in compliance with ISO 9001, ISO 13485.
Envase: Packed 10 boxes per case.
Precaución: Keep storage area cool, dry and dust free, avoid ventilation and storage close to photocopy equipment. Copper ions discolour the glove. Protect gloves against ultraviolet light sources, as sunlight and oxidizing agents. Storage above 38 °C (100 °F) will lead to accelerated aging and should be avoided under any circumstances. Long term storage in bulk can lead to pleats, stickiness, and early aging of the glove and should be avoided.