Compact Switching Power Supply, Selectable Output 3 to 12 V, Digi-Key
Supplier: Digi-Key
1528-1448AD-ND
MSPP-15281448AEA 23 USD
MSPP-15281448A
Compact Switching Power Supply, Selectable Output 3 to 12 V, Digi-Key
Power Supplies
Its a switching wall adapter where the voltage can be easily changed. This model is 10 Watts and has 6 selectable outputs ranging from 3 to 12 V and up to 1.5 A output. It has a 2-prong plug, but takes 110 to 240 V so you can use it anywhere with a simple plug adapter.
  • Multi changeable plug DC-cord
  • Universal AC input plug available
  • Over load, over current, over heat protection
  • Extra low consumption during standby (about 8% of linear adapter)
It comes with a range of tips, such as 2.5 mm mono, 3.5 mm mono, 5.5 mm/2.5 mm, 2.5 mm/2.1 mm, 4 mm, etc. You can also change it from center positive (most common) to center negative.
Certificaciones: International approval CE EN60950, UL1950, CSA22.2 No.950, EMC approval EN55022.