United States
Order Entry
NotI
NotI
 
undefined
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Application:
  • Source:
  • Environmentally Preferable:

 

 

  •  
     
    Concentration
    Size
    Supplier no.
     
     
  • NotI
    NotI
    101228-962
    R0189S
    10000 units/ml
    500 units
    R0189S
       
       
    Each
    Each - $85.00
    $85.00
     
    Each
       $85.00

     
    Small Business Enterprise
    Small Business Enterprise
  • NotI
    NotI
    101228-958
    R0189L
    10000 units/ml
    2500 units
    R0189L
       
       
    Each
    Each - $354.00
    $354.00
     
    Each
       $354.00

     
    Small Business Enterprise
    Small Business Enterprise