Biomek® 4000 Automated Workstation for Nucleic Acid Purification, Beckman Coulter
Supplier: Beckman Coulter
Biomek®
B22637 B22640 B22638
75816-480EA 101876.46 USD
75816-480 75816-482 75816-486
Biomek® 4000 Automated Workstation for Nucleic Acid Purification, Beckman Coulter
Automated Nucleic Acid Purification Systems
A compact, nonsterile, semi-automated liquid handler ideal for smaller laboratories. Standardize daily pipetting routines, maintain sample quality, and generate repeatable, reliable results.
  • Ideal for low-throughput workflows
  • 12 Deck positions
  • Flexible pipetting with interchangeable pipetting tools and gripper
  • Fits in laminar flow hood
  • Optional enclosure to protect samples from airborne particles
The Biomek® 4000 Automated Workstation features liquid-level sensing, windows 7 operating system, and a 22" display control screen. Execute accurate intra-well pipetting, efficient wash, and bulk dispense tool. Supports labware from tubes to 384-well microplates. Biomek 4000 has a dynamic volume pipetting range of 1 to 1000 µl for precise and robust pipetting, available in both single- and 8-channel.
Simple system operation is powered by icon-based software with data tracking and LIMS connectivity, guided system setup, factory-calibrated pipetting tools, and automated deck framing.
Application focused solutions includes consultative workflow analysis, expert application and technical support, and automated demonstration methods and capabilities for genomic, drug discovery, and basic research workflows.
Suitable software includes Biomek® software, SAMI EX scheduling software, Bbiomek® powerpack software, Biomek® method launcher, or DART 2.0 software.