InnuPure C16 touch, Analytik Jena

Supplier: Analytik Jena
InnuPure
845-00020-2
76414-004EA 30959.37 USD
76414-004
InnuPure C16 touch, Analytik Jena
Automated Nucleic Acid Purification Systems
Whether magnetic particle separation or novel SmartExtraction technology – the InnnuPure C16 touch significantly increases efficiency in nucleic acid extraction. Whatever your extraction method, the InnuPure C16 touch not only delivers higher yields and highly reproducible results but also helps you find the ideal combination of kit, protocol, and parameters.

  • Highly flexible DNA and RNA extraction
  • Two reliable functions: magnetic particle separation and SmartExtraction
  • The best solution for every material
  • Adapts to your requirements

The InnuPure C16 touch combines high-precision liquid handling and the automated extraction of high-quality nucleic acids. The device sets new standards in terms of safety and user-friendliness. Pipetting errors are not an issue thanks to ready-to-use reagent plastic.
• Pipette tips with aerosol filters prevent contamination
• Optional UV lamp prevents further sources of contamination
• Intuitive operation with a 10"-tablet PC
•Software with individual optimization recommendations, depending on the materials involved

Weight: Approx. 30 kg
Dimension (W x H x L): 400 mm x 543 mm x 550 mm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR