Avantor® Hichrom HI-BIODIESEL 103 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI39-32-025-30
76426-550EA 1304.05 USD
76426-550
Avantor® Hichrom HI-BIODIESEL 103 GC Columns
Chromatography Columns
Application specifc column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.