Volatile Organics Mix with 76 Certified Components for US EPA Method 8260 and CLP Series Methods
Supplier: Avantor
VWR Chemicals BDH®
BDH0707-1ML
BDH0707-1MLEA 448.34 USD
BDH0707-1ML
Volatile Organics Mix with 76 Certified Components for US EPA Method 8260 and CLP Series Methods
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.