Purgeable Aromatics for Gasoline Identification for US EPA Method 602
Supplier: Avantor
VWR Chemicals BDH®
BDH0793-1ML
BDH0793-1MLEA 45.24 USD
BDH0793-1ML
Purgeable Aromatics for Gasoline Identification for US EPA Method 602
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 602 is a general analytical method for the monitoring of halogenated aromatic compounds in municipal and industrial wastewater by GC/PID.