Phenolics Mix B for US EPA Method 8041
Supplier: Avantor
VWR Chemicals BDH®
BDH0761-1ML
BDH0761-1MLEA 79.2 USD
BDH0761-1ML
Phenolics Mix B for US EPA Method 8041
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8041 is a general analytical method for the monitoring of phenols for GC/FID or GC/ECD.