Nitrosamines Mix for US EPA Method 8270
Supplier: VWR Chemicals BDH®
BDH0769-1ML
BDH0769-1MLEA 122.14 USD
BDH0769-1ML
Nitrosamines Mix for US EPA Method 8270
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.