Long List Appendix IX Compound Mix E for US EPA Method 8260B and CLP Series Methods, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0765-1ML
BDH0765-1MLEA 48.86 USD
BDH0765-1ML
Long List Appendix IX Compound Mix E for US EPA Method 8260B and CLP Series Methods, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Volatiles standard.
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.
CLP Series Methods - The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846.
The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.