High Concentration Base/Neutral Surrogate Standards for CLP Series Methods
Supplier: VWR Chemicals BDH®
BDH0783-5ML
BDH0783-5MLEA 172.77 USD
BDH0783-5ML
High Concentration Base/Neutral Surrogate Standards for CLP Series Methods
Standards Environmental Analysis Standards
Semivolatiles standard.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.