Free Acids Analyte Mix for US EPA Method 552
Supplier: VWR Chemicals BDH®
BDH0752-1ML
BDH0752-1MLEA 98.32 USD
BDH0752-1ML
Free Acids Analyte Mix for US EPA Method 552
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 552 is an analytical method for the monitoring of haloacetic acids and phenolic disinfectant by-products in drinking water by GC/ECD.