Balance Mix, Option A, for US EPA Method 8270 and CLP Series Methods
Supplier: VWR Chemicals BDH®
BDH0800-1ML
BDH0800-1MLEA 45.29 USD
BDH0800-1ML
Balance Mix, Option A, for US EPA Method 8270 and CLP Series Methods
Standards Environmental Analysis Standards
Semivolatiles standard.
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.