Alternate Surrogate Standard for US EPA Method 8260, VWR Chemicals BDH®
Supplier: VWR Chemicals BDH®
BDH0764-1ML
BDH0764-1MLEA 50.05 USD
BDH0764-1ML
Alternate Surrogate Standard for US EPA Method 8260, VWR Chemicals BDH®
Standards Environmental Analysis Standards
Volatiles standard
US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.