3-Ply Level 1 Face Masks with Earloops, Oakridge

MASK-LVL1-GEN
76415-020CS 442.8 USD
76415-020
3-Ply Level 1 Face Masks with Earloops, Oakridge
Masks Surgical Masks
This mask conforms to any size or shape face. Custom fit is achieved by pinching the aluminum nose strip.

  • Fluid resistance: 120 mm Hg
  • BFE >98% at 3.0 µm
  • PFE >95% at 0.1 µm
  • Delta P (breathability) <4.0 H₂O/cm²
  • Flame spread class 1
  • Extra long, aluminum nose piece

Certificaciones: FDA registered product code QKR - Device Class I ear loop mask. Meets EN 14683 Rating - Type IIR Standard.

Envase: Masks are packaged 50 per box, 2000 per case.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR