Axygen® Mini Plate Spinner Centrifuge, Corning
Supplier: Corning
PLATESPINNER-230EU PLATESPINNER-120
76417-686CS 872.08 USD
76417-686 76417-688
Axygen® Mini Plate Spinner Centrifuge, Corning
Centrifuges Benchtop Centrifuges
Axygen® Axyspin plate spinner is designed specifically for quick and easy centrifugation of samples in PCR plates or microplates.
  • Accepts skirted and non-skirted and all standard PCR plates
  • Exceptionally small footprint
  • Capacity for two standard plates
  • Top loading design
  • Less than ¼ the size of most plate centrifuges
  • Starts and stops in seconds
  • Quickly spin down droplets and condensation
Operating range: 4 to 35 °C