Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Chemical Resistant Sleeve Protectors

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"9"
Sort by:
 
 
 
 

Supplier:  Kimberly-Clark
Description:   Sleeve protectors are manufactured from breathable, microporous film laminate, which is an excellent particle barrier (100% dry particle holdout betwe...
Product and/or packaging is recyclable or reduces waste
Supplier:  Honeywell Safety Products
Description:   Sleeves feature a five-layer fabric compacted into 2.7mil
Catalog Number: (13450-338)
Supplier:  DuPont
Description:   A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
Supplier:  Kimberly-Clark
Description:   The KLEENGUARD® A80 line of apparel is designed to provide excellent worker protection in chemical handling/mixing/cleanup, hazardous material handlin...
Supplier:  Kimberly-Clark
Description:   Patented three-layer fabric construction features a middle layer of microporous film that allows heat and sweat vapor to escape while protecting again...
Product and/or packaging is recyclable or reduces waste
Supplier:  Kimberly-Clark
Description:   The KLEENGUARD® A70 line of apparel is designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, pe...
Supplier:  Ansell Healthcare
Description:   Embossed polyethylene sleeves provide excellent protection against chemicals, snags, punctures, and abrasions.
Catalog Number: (490008-104)
Supplier:  Microflex
Description:   This embossed polyethylene sleeve provides excellent protection against chemicals, snags, punctures, and abrasions.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next